missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 6B Polyclonal Antibody

Descripción
Cytokeratin 6B Polyclonal Antibody for Western Blot, ELISA
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. As many as six of this type II cytokeratin (KRT6) have been identified; the multiplicity of the genes is attributed to successive gene duplication events. The genes are expressed with family members KRT16 and/or KRT17 in the filiform papillae of the tongue, the stratified epithelial lining of oral mucosa and esophagus, the outer root sheath of hair follicles, and the glandular epithelia. Mutations in these genes have been associated with pachyonychia congenita. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.
Especificaciones
Especificaciones
| Antígeno | Cytokeratin 6B |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.09 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | KRT6B |
| N.º de referencia del gen | P04259 |
| Alias de gen | BB144589; CK6B; CK-6B; cytokeratin 6B; cytokeratin-6B; K6B; keratin 6B; keratin 6B, type II; keratin complex 2, basic, gene 6b; keratin, epidermal, type II, K6B; keratin, type II cytoskeletal 6B; keratin-6B; keratin-6-beta; keratin-like 1 (a type II keratin sequence); Krt2-6b; KRT6B; KRTL1; mK6[b]; mK6-beta; PC2; PC4; type-II keratin Kb10 |
| Símbolos de los genes | KRT6B |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?