Learn More
Cytokeratin 18 Mouse anti-Human, Mouse, Rat, Clone: 1G11C4, Proteintech
Código de producto. 16847835 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16847835 20 μL 20 microlitros
16837835 150 μL 150 microlitros
2 options
Código de producto. 16847835
Marca: Proteintech 661871IG20UL
Mouse Monoclonal Antibody
Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial maligcies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
Especificaciones
Antígeno Cytokeratin 18
Aplicaciones Immunocytochemistry, Flow Cytometry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 1G11C4
Concentración 0.87 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica KRT18
N.º de referencia del gen P05783, P05784, Q5BJY9
Alias de gen CK 18, CYK18, Cytokeratin 18, K18, keratin 18, KRT18
Símbolos de los genes KRT18
Especie del huésped Mouse
Inmunógeno Cytokeratin 18 Fusion Protein Ag1260
Método de purificación Protein G
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16668, 294853, 3875
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1