missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 15 Mouse anti-Human, Mouse, Clone: 6B4F8, Proteintech

Product Code. 16847285 Shop All Proteintech Products
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Unit Size:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16847285 20 μL 20 microlitros
16837285 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16847285 Supplier Proteintech Supplier No. 602471IG20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21.2.

Specifications

Antígeno Cytokeratin 15
Aplicaciones Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 6B4F8
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica KRT15
N.º de referencia del gen P19012, Q61414
Alias de gen CK 15, CK15, Cytokeratin 15, K15, K1CO, keratin 15, KRT15, KRTB
Símbolos de los genes KRT15
Especie del huésped Mouse
Inmunógeno Cytokeratin 15 Fusion Protein Ag0185
Método de purificación Protein G
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16665, 3866
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.