missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16855-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is specifically against KRT10.
Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (i.e.., skin) and non-keratinizing cells (i.e.. mesothelial cells). Although not a traditional marker for endothelial cells, cytokeratins have also been found in some microvascular endothelial cells. At least 20 different cytokeratins (CK) in the molecular range of 40-70 kDa and isoelectric points of 5-8.5 can be identified using two dimensional gel electrophoresis. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells. Monoclonal antibodies to cytokeratin proteins can be useful markers for tumor identification and classification.Especificaciones
| Cytokeratin 10 | |
| Polyclonal | |
| Unconjugated | |
| KRT10 | |
| CK 10, CK10, Cytokeratin 10, K10, keratin 10, KPP, KRT10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16661, 3858, 450225 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02535, P13645, Q6IFW6 | |
| KRT10 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido