missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytohesin 1 Monoclonal Antibody (2E11)

Descripción
MA1-060 detects cytohesin-1 from human and mouse cells as well as recombinant human protein. MA1-060 has been successfully used in Western blot and IF procedures. By Western blot, this antibody detects an ~48 kDa protein representing cytohesin-1. The MA1-060 immunogen is purified, His-tagged cytohesin-1.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
Especificaciones
Especificaciones
| Antígeno | Cytohesin 1 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 2E11 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1mg/mL BSA and 0.05% sodium azide |
| génica | CYTH1 |
| N.º de referencia del gen | Q15438, Q9QX11 |
| Alias de gen | B2-1; CLM1; CTH-1; CYTH1; CYTIP; cytoadhesin 1; cytohesin 1; CYTOHESIN-1; D17S811E; FLJ34050; FLJ41900; homolog of secretory protein SEC7; mKIAA4240; mSec7-1; PH, SEC7 and coiled-coil domain-containing protein 1; pleckstrin homology, Sec7 and coiled/coil domains 1(cytohesin 1); pleckstrin homology, Sec7 and coiled-coil domains 1; Pscd1; rSec7-1; SEC7; SEC7 homolog A; SEC7 homolog B2-1; Sec7a |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?