missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15962-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes and plays a role in epidermal development and maintece. Stefins have been proposed as prognostic and diagnostic tools for cancer.Especificaciones
| Cystatin A | |
| Polyclonal | |
| Unconjugated | |
| CSTA | |
| CSTA, Cystatin A, cystatin A (stefin A), Cystatin AS, Stefin A, STF1, STFA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1475, 209294 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01040, P56567 | |
| Csta | |
| Cystatin A Fusion Protein Ag8733 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?