missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CYR61 is a secreted heparin binding protein, encoded by a growth factor inducible immediate-early gene that associates with the extracellular matrix and connective tissue. CYR61 is a member of a distinct family of angiogenic and vasculogenic regulators designated CCN proteins, which includes connective tissue growth factor (CTGF) and the mouse CYR61 homolog, Fisp12. As an angiogenic inducer, CYR61 binds to the cell surface receptor integrin alphavbeta3, where it then stimulates cell adhesion and migration and promotes DNA synthesis of human vascular endothelial cells. Expression of CYR61 is elevated during vessel growth, wound healing and chondrocyte differentiation. CYR61 is also detected in a wide variety of tumors as it induces tumor growth and functions as a marker of tumor progression.
Especificaciones
Especificaciones
| Antígeno | CYR61/CCN1 |
| Aplicaciones | Immunofluorescence, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.33 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | CYR61 |
| N.º de referencia del gen | O00622 |
| Alias de gen | CCN family member 1, CCN1, CYR61, GIG1, IBP 10, IGF binding protein 10, IGFBP 10, IGFBP10, Protein CYR61, Protein GIG1 |
| Símbolos de los genes | CYR61 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?