missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP2D6 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 16300725
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
16300725 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16300725

Marca: Invitrogen™ PA583766

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: EYDDPRFLRL LDLAQEGLKE ESGFLREVLN AVPVLLHIPA LAGKVLRFQK AFLTQLDELL TEHRMTWDPA QPPRDLTE Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 65%.

Cytochrome p450 2D6 (CYP2D6) is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize as many as 20% of commonly prescribed drugs. Its substrates include debrisoquine, an adrenergic-blocking drug; sparteine and propafenone, both anti-arrythmic drugs; and amitryptiline, an anti-depressant. The gene is highly polymorphic in the population; certain alleles result in the poor metabolizer phenotype, characterized by a decreased ability to metabolize the enzyme's substrates. The gene is located near two cytochrome P450 pseudogenes on chromosome 22q13.1. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CYP2D6
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.20 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica CYP2D6
N.º de referencia del gen P10635
Alias de gen AI303445; Cholesterol 25-hydroxylase; CPD6; CYP2D; Cyp2d10; Cyp2d-10; Cyp2d13; Cyp2d18; Cyp2d-18; Cyp2d22; Cyp2d3; Cyp2d-3; Cyp2d4; Cyp2d-4; Cyp2d4v1; Cyp2d4v2; Cyp2d6; CYP2D7AP; CYP2D7BP; CYP2D7P2; CYP2D8P2; CYP2DL1; CYPIID10; CYPIID18; CYPIID3; CYPIID4; CYPIID6; Cytochrome P450 2D10; cytochrome P450 2d18; Cytochrome P450 2D-29; Cytochrome P450 2D3; cytochrome P450 2D-35; cytochrome P450 2D4; Cytochrome P450 2D6; cytochrome P450 family 2 subfamily D member 6; cytochrome P450, 2d10; cytochrome P450, family 2, subfamily d, polypeptide 10; cytochrome P450, family 2, subfamily d, polypeptide 13; cytochrome P450, family 2, subfamily d, polypeptide 22; cytochrome P450, family 2, subfamily d, polypeptide 3; cytochrome P450, family 2, subfamily d, polypeptide 4; cytochrome P450, family 2, subfamily d, polypeptide 6; cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2; cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2; cytochrome P450, subfamily 2D, polypeptide 6; cytochrome P450, subfamily II (debrisoquine, sparteine, etc., -metabolising), polypeptide 7 pseudogene 2; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolising), polypeptide 8 pseudogene 2; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing), polypeptide 6; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing)-like 1; cytochrome P450, subfamily IID, polypeptide 6; Cytochrome P450, subfamily IID3; Cytochrome P450, subfamily IID4; cytochrome P450-16-alpha; Cytochrome P450CB; Cytochrome P450-CMF3; cytochrome P450-DB1; Cytochrome P450-DB3; cytochrome P450-DB4; debrisoquine 4-hydroxylase; flavoprotein-linked monooxygenase; microsomal monooxygenase; nonfunctional cytochrome P450 2D6; P450 2D-29/2D-35; P450-2D; P450C2D; P450-CMF3; P450DB1; P450-DB1; P450-DB3; P450-DB4; testosterone 16-alpha hydroxylase; xenobiotic monooxygenase
Símbolos de los genes CYP2D6
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human CYP2D6. Recombinant protein control fragment (Product #RP-100044).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1565
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.