Learn More
Invitrogen™ CYP2C9 Polyclonal Antibody

Descripción
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Pig and Horse.
Especificaciones
Especificaciones
| Antígeno | CYP2C9 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 |
| génica | CYP2C9 |
| N.º de referencia del gen | P11712 |
| Alias de gen | (R)-limonene 6-monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; Cholesterol 25-hydroxylase; CPC9; CYP2C; CYP2C10; CYP2C9; CYPIIC9; Cytochrome P450 2C9; cytochrome P450 family 2 subfamily C member 9; Cytochrome P450 MP-4; Cytochrome P450 MP-8; Cytochrome P450 PB-1; cytochrome P-450 S-mephenytoin 4-hydroxylase; cytochrome P450, family 2, subfamily C, polypeptide 9; cytochrome P-450MP; flavoprotein-linked monooxygenase; MGC149605; MGC88320; microsomal monooxygenase; P450IIC9; S-mephenytoin 4-hydroxylase; xenobiotic monooxygenase |
| Símbolos de los genes | CYP2C9 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.