missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP2C19 Polyclonal Antibody

Description
CYP2C19 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF
CYP2C19 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize many xenobiotics, including the anticonvulsive drug mephenytoin, omeprazole, diazepam and some barbiturates. Polymorphism within its gene is associated with variable ability to metabolize mephenytoin, known as the poor metabolizer and extensive metabolizer phenotypes.
Specifications
Specifications
| Antígeno | CYP2C19 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 2.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | CYP2C19 |
| N.º de referencia del gen | P33261 |
| Alias de gen | (R)-limonene 6-monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; CPCJ; CYP2C; CYP2C19; CYPIIC17; CYPIIC19; cyto; cytochrome P450 2C19; cytochrome P450 family 2 subfamily C member 19; cytochrome P-450 II C; cytochrome P450, family 2, subfamily C, polypeptide 19; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 19; cytochrome P450-11A; Cytochrome P450-254C; Fenbendazole monooxygenase (4'-hydroxylating); flavoprotein-linked monooxygenase; mephenytoin 4'-hydroxylase; mephenytoin 4-hydroxylase; microsomal monooxygenase; P450-11A; P450-254C; P450C2C; P450IIC19; S-mephenytoin 4-hydroxylase; xenobiotic monooxygenase |
| Símbolos de los genes | CYP2C19 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?