Learn More
CYP26B1 Mouse anti-Human, Clone: OTI4A8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF811497
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and the synthesis of cholesterol, steroids and other lipids. The enzyme encoded by this gene is involved in the specific inactivation of all-trans-retinoic acid to hydroxylated forms, such as 4-oxo-, 4-OH-, and 18-OH-all-trans-retinoic acid.Especificaciones
| CYP26B1 | |
| Monoclonal | |
| Unconjugated | |
| CYP26B1 | |
| CYP26A2, P450RAI-2, P450RAI2, RHFCA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56603 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4A8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NR63 | |
| CYP26B1 | |
| Human recombit protein fragment corresponding to amino acids 229-512 of human CYP26B1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.