Learn More
Invitrogen™ Cyclin L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA543615
Descripción
Peptide sequence: TGQVLFQRFF YTKSFVKHSM EHVSMACVHL ASKIEEAPRR IRDVINVFHR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
CCNL2 belongs to the cyclin family, Cyclin L subfamily. CCNL2 is a transcriptional regulator which participates in regulating the pre-mRNA splicing process. CCNL2 also modulates the expression of critical apoptotic factor, leading to cell apoptosis.
Especificaciones
| Cyclin L2 | |
| Polyclonal | |
| Unconjugated | |
| CCNL2 | |
| 1700010A01Rik; 1810019L15Rik; 2010319M22Rik; AA409936; Ania6b; ANIA-6B; AW261738; Ccnl2; CCNM; CCNS; Cyclin Ania-6b; cyclin L2; cyclin M; cyclin S; cyclin-L2; HCLA-150; HCLA-ISO; HLA-ISO; hypothetical protein LOC538796; MNCb-5160; paneth cell enhanced expression; paneth cell-enhanced expression protein; PCEE; SB138 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 81669 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96S94 | |
| CCNL2 | |
| Synthetic peptide directed towards the N-terminal of human CCNL2 (aa 101-150). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.