missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin B1 Recombinant Rabbit Monoclonal Antibody (1Q3E9)

Descripción
Immunogen sequence: LRVTRNSKIN AENKAKINMA GAKRVPTAPA ATSKPGLRPR TALGDIGNKV SEQLQAKMPM KKEAKPSATG KVIDKKLPKP LEKVPMLVPV PVSEPVPEPE PEPEPEPVKE EKLSPEPILV DTASPSPMET SGCAPAEEDL CQAFSDVILA VNDVDAEDGA DPNLCSEYVK DIYAY.
Cyclins bind to and regulate the activity of the Cyclin dependent protein kinases (CDKs). Cyclin B1 or CCNB1 is a regulatory protein involved in mitosis. It is essential for the control of the cell cycle at the G2/M transition. Cyclin B1 complexes with p34 (cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. Cyclin B1 is not ubiquitinated during G2/M phase, resulting in its steady accumulation during G2 phase, followed by abrupt APC dependent destruction at the end of mitosis. Destruction of Cyclin B1 is required for cell cycle progression. Cyclin B1 is overexpressed in various cancers, including breast, prostate, and non-small cell lung cancer.
Especificaciones
Especificaciones
| Antígeno | Cyclin B1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Recombinant Monoclonal |
| Clon | 1Q3E9 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | Ccnb1 |
| N.º de referencia del gen | P14635, P24860 |
| Alias de gen | Ccn-2; CCNB; Ccnb1; Ccnb1-rs1; Ccnb1-rs13; Cycb; Cycb1; Cycb1-rs1; Cycb-4; Cycb-5; cyclin B; cyclin B1; cyclin B1, related sequence 1; cyclin B1, related sequence 13; G2/mitotic-specific cyclin B1; G2/mitotic-specific cyclin-B1; I79_013589; OTTHUMP00000221981; OTTHUMP00000223023 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?