missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclin B1 Monoclonal Antibody (V152)

Código de producto. 15336281
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
15336281 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15336281

Marca: Invitrogen™ MA1155

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA1-155 detects Cyclin B1 in mammalian samples. MA1-155 has been successfully used in Immunohistochemistry (paraffin), Immunofluorescence, Immunoprecipitation, Flow Cytometry and Western Blot procedures. Immunoprecipitation and Western Blot analysis with MA1-155 show the accumulation of a prominent band at ~52 kDa in camptothecin and hydroxyurea treated cells. MA1-155 also detects additional unknown bands at ~35 and ~75 kDa. In Immunofluorescence applications, MA1-155 shows cyclin B1 co-localization with microtubules, whereas MA1-156 shows cyclin B2 staining consistent with the Golgi region. MA1-155 specifically detects Cyclin B1 in in whole cell lysates, but will also cross-react with purified recombinant human Cyclin B2 in Western Blot applications.

Cyclins bind to and regulate the activity of the Cyclin dependent protein kinases (CDKs). Cyclin B1 or CCNB1 is a regulatory protein involved in mitosis. It is essential for the control of the cell cycle at the G2/M transition. Cyclin B1 complexes with p34 (cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. Cyclin B1 is not ubiquitinated during G2/M phase, resulting in its steady accumulation during G2 phase, followed by abrupt APC dependent destruction at the end of mitosis. Destruction of Cyclin B1 is required for cell cycle progression. Cyclin B1 is overexpressed in various cancers, including breast, prostate, and non-small cell lung cancer.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Cyclin B1
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon V152
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
génica Ccnb1
N.º de referencia del gen P14635, P24860, P30277, Q08301
Alias de gen Ccn-2; CCNB; Ccnb1; Ccnb1-rs1; Ccnb1-rs13; Cycb; Cycb1; Cycb1-rs1; Cycb-4; Cycb-5; cyclin B; cyclin B1; cyclin B1, related sequence 1; cyclin B1, related sequence 13; G2/mitotic-specific cyclin B1; G2/mitotic-specific cyclin-B1; I79_013589; OTTHUMP00000221981; OTTHUMP00000223023
Símbolos de los genes Ccnb1
Especie del huésped Mouse
Inmunógeno Hamster Cyclin B1.
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100770842, 25203, 268697, 891
Especies diana Human, Mouse, Rat, Monkey, Hamster
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.