missing translation for 'onlineSavingsMsg'
Learn More

CXCR6 (CD186) Mouse anti-Human, Brilliant Violet 510, Clone: 13B 1E5, BD Optibuild™

Código de producto. 15818379
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15818379 50 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15818379 Proveedor BD Biosciences N.º de proveedor 743598

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The 13B 1E5 monoclonal antibody specifically binds to C-X-C chemokine receptor type 6 (CXCR6), which is also known as CD186, BONZO, T-lymphocyte-expressed seven-transmembrane domain receptor (TYMSTR), and Seven transmembrane receptor-like from clone 33 (STRL33). CXCR6 is a G-protein coupled chemokine receptor that is expressed on subsets of activated and memory T cells and NKT cells. CXCR6 binds to soluble CXCL16 and membrane-anchored CXCL16 expressed by dendritic cells and macrophages. The CXCL16 and CXCR6 interaction activates Akt and mTor signaling. This regulates cellular migration including the recruitment of tumor-infiltrating lymphocytes and may contribute to the progression and metastasis of various cancers. CXCR6 also can serve as a coreceptor for certain strains of HIV-1 and HIV-2.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CXCR6 (CD186)
Clasificación Monoclonal
Clon 13B 1E5
Concentración 0.2mg/mL
Conjugado Brilliant Violet 510
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Alias de gen C-X-C chemokine receptor type 6, BONZO, STRL33, TYMSTR
Especie del huésped Mouse
Inmunógeno Human CXCR6
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.