missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CXCR4 (CD184) Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE

Código de producto. p-200111873
Change view
Click to view available options
Cantidad:
1 mg
100 μg
Tamaño de la unidad:
1 miligramo
Código de producto. Cantidad unitSize
30284415 100 μg -
30282578 1 mg 1 miligramo
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30284415

Marca: Invitrogen™ MA559345

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Dana-Farber patent anti-CXCR4.

CXCR4 (Fusin, LESTR, HUMSTR) is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a co-receptor for X4 HIV-1 entry into CD4+ cells. CXCR4 has been proved to be the co-receptor for HIV-2's binding to CD4 through envelope glycoprotein gp 120 and promotes Env-mediated fusion of the virus. In other cases, CXCR4 can even function as the only receptor for HIV-2's binding to the CD4 host cells. CXCR4 is the receptor for the C-X-C chemokine CXCL12/SDF-1 that transduces a signal by increasing intracellular calcium ion levels and enhancing MAPK1/MAPK3 activation. CXCR4 acts as a receptor for extracellular ubiquitin; leading to enhanced intracellular calcium ions and reduced cellular cAMP levels. Also, CXCR4 is involved in hematopoiesis and in cardiac ventricular septum formation, and plays an essential role in vascularization of the gastrointestinal tract, probably by regulating vascular branching and/or remodeling processes in endothelial cells. In the CNS, CXCR4 could mediate hippocampal-neuron survival and is involved in cerebellar development. CXCR4 binds bacterial lipopolysaccharide (LPS) and mediates LPS-induced inflammatory response, including TNF secretion by monocytes. Antibodies to CXCR4 block HIV-1 and HIV-2 fusion and infection of human target cells. The amino-terminal domain and the second extracellular loop of CXCR4 serve as HIV biding sites.
TRUSTED_SUSTAINABILITY

Specifikationer

Antígeno CXCR4 (CD184) Biosimilar
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Clasificación Recombinant Monoclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Especie del huésped Human
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.