missing translation for 'onlineSavingsMsg'
Learn More

PeproTech CXCL9 Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. p-7245549
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
Código de producto. Cantidad unitSize
17824873 50 μg 50 microgramos
17845313 100 μg 100 microgramos
17865283 1 mg 1 miligramo
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17824873

Marca: PeproTech 500P5050UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: TPVVRKGRCS CISTNQGTIH LQSLKDLKQF APSPSCEKIE IIATLKNGVQ TCLNPDSADV KELIKKWEKQ VSQKKKQKNG KKHQKKKVLK VRKSQRSRQK KTT. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MIG (CXCL9). Anti-Human MIG (CXCL9)-specific antibody was purified by affinity chromatography employing an immobilized Human MIG (CXCL9) matrix. Sandwich ELISA: To detect Human MIG (CXCL9) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human MIG (CXCL9) (500-P50Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human MIG (CXCL9). Western Blot: To detect Human MIG (CXCL9) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human MIG (CXCL9) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

MIG, also known as CXCL9, is a T-cell chemoattractant inducible by gamma interferon that is a member of the CXC chemokine family of cytokines.This gene is mapped to 4q21. It is noted that, although the best-described activities of the chemokines are as chemotactic factors, chemokines also have an effect on T-cell activation, angiogenesis, and HIV infection. While most CXC chemokines are chemotactic for neutrophils, MIG and INP10 are unusual and similar in being CXC chemokines that are chemotactic for lymphocytes and inactive in neutrophils.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CXCL9
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica CXCL9
N.º de referencia del gen Q07325
Alias de gen BB139920; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CMK; crg-10; CXC; CXC motif chemokine; C-X-C motif chemokine 9; C-X-C motif chemokine ligand 9; CXCL; CXCL9; gamma interferon-induced monokine; gamma-interferon-induced monokine; HuMIG; M119; Mig; monokine induced by gamma interferon; monokine induced by interferon gamma; monokine induced by interferon-gamma; MuMIG; protein m119; SCYB9; small inducible cytokine B subfamily (Cys-X-Cys), member 9; small inducible cytokine B9; small-inducible cytokine B9
Símbolos de los genes CXCL9
Especie del huésped Rabbit
Inmunógeno E.coli-derived Recombinant Human MIG (CXCL9).
Método de purificación Antigen affinity chromatography
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4283
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.