missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL16 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA579108
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The synthetic peptide sequence is 103-116aa, HGKSFHHQKHLPQA Add 0.2 mL of distilled water, will yield a concentration of 500 μg/mL.
Acts as a scavenger receptor on macrophages, which specifically binds to OxLDL (oxidized low density lipoprotein), suggesting that it may be involved in pathophysiology such as atherogenesis. Induces a strong chemotactic response. Induces calcium mobilization. Binds to CXCR6/Bonzo.
Especificaciones
| CXCL16 | |
| Polyclonal | |
| PBS with 5MG BSA, 5MG BSA and 0.05MG sodium azide, 0.05MG sodium azide, 0.05MG thimerosal, 0.05MG thimerosal | |
| Q8BSU2 | |
| Cxcl16 | |
| A synthetic peptide corresponding to a sequence in the middle region of mouse CXCL16. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Cxcl16 | |
| C-X-C motif chemokine 16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SR-PSOX, Small-inducible cytokine B16, Transmembrane chemokine CXCL16, Cxcl16, Srpsox | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 66102 | |
| -20°C | |
| Lyophilized |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido