missing translation for 'onlineSavingsMsg'
Learn More

PeproTech CXCL14 Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. 17825413
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
Código de producto. Cantidad unitSize
17825413 1 mg 1 miligramo
17815683 50 μg 50 microgramos
17845273 100 μg 100 microgramos
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17825413

Marca: PeproTech 500P2371MG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: SKCKCSRKGP KIRYSDVKKL EMKPKYPHCE EKMVIITTKS VSRYRGQEHC LHPKLQSTKR FIKWYNAWNE KRRVYEE. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BRAK (CXCL14). Anti-Human BRAK (CXCL14)-specific antibody was purified by affinity chromatography employing an immobilized Human BRAK (CXCL14) matrix. Sandwich ELISA: To detect Human BRAK (CXCL14) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BRAK (CXCL14) (500-P237Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BRAK (CXCL14). Western Blot: To detect Human BRAK (CXCL14) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BRAK (CXCL14) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

This gene belongs to the cytokine gene family which encode secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC (Cys-X-Cys) subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes but not for lymphocytes, dendritic cells, neutrophils or macrophages. It has been implicated that this cytokine is involved in the homeostasis of monocyte-derived macrophages rather than in inflammation.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CXCL14
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica Cxcl14
N.º de referencia del gen O95715
Alias de gen 1110031L23Rik; 1200006I23Rik; AI414372; B-cell and monocyte-activating chemokine; BMAC; bolekine; BRAK; breast and kidney; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 14; chemokine BRAK; CXC; CXC chemokine in breast and kidney; CXC motif chemokine; C-X-C motif chemokine 14; C-X-C motif chemokine ligand 14; CXCL; Cxcl14; hypothetical protein LOC511771; Kec; Kidney-expressed chemokine CXC; KS1; MGC10687; MIP-2 gamma; MIP2G; MIP-2g; MIP2gamma; musculus CXC chemokine MIP-2gamma; NJAC; PSEC0212; SCYB14; small inducible cytokine B14; small inducible cytokine subfamily B (Cys-X-Cys), member 14; small inducible cytokine subfamily B (Cys-X-Cys), member 14 (BRAK); small-inducible cytokine B14; tumor-suppressing chemokine; UNQ240/PRO273
Símbolos de los genes Cxcl14
Especie del huésped Rabbit
Inmunógeno E.coli-derived Recombinant Human BRAK (CXCL14).
Método de purificación Antigen affinity chromatography
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9547
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.