Learn More
PeproTech CXCL12 alpha (SDF-1 alpha) Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Descripción
AA Sequence of recombinant protein: KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human SDF-1alpha (CXCL12). Anti-Human SDF-1alpha (CXCL12)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human SDF-1alpha by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human SDF-1alpha (500-P87A) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human SDF-1alpha. Western Blot: To detect Human SDF-1alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human SDF-1alpha is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P87ABT-1MG will be provided as 2 x 500 μg
Especificaciones
Especificaciones
| Antígeno | CXCL12 alpha (SDF-1 alpha) |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1-1.0 mg/mL |
| Conjugado | Biotin |
| Formulación | PBS with no preservative |
| génica | CXCL12 |
| N.º de referencia del gen | P48061 |
| Alias de gen | 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; Cxcl12; H-SDF-1-alpha; Intercrine reduced in hepatomas; IRH; Pbsf; pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SCYB12; SDF 1 a; SDF 1 alpha; SDF 1α; SDF1; SDF-1; SDF1 alpha; SDF-1 gamma; SDF1A; SDF-1-alpha(3-67); SDF1B; SDF-1-beta(3-72); SDF1α; Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tpar1 |
| Símbolos de los genes | CXCL12 |
| Mostrar más |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.