missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA533032
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CTR1 Polyclonal Antibody for IHC (P)
The trace metal copper (Cu) plays a crucial role in mammalian cells as a cofactor for many enzymes. Cu-related genetic diseases, such as Menkes disease and Wilson disease, arise from a lack of Cu homeostasis in mammalian cells. CTR1 is a high-affinity copper-uptake protein. The C-terminal domain is similar to the Raf family of protein kinases, but it's first two-thirds encodes a novel protein domain. CTR1 provides the primary avenue for copper uptake in mammalian cells, thereby, affecting Cu homeostasis and embryonic development.
Especificaciones
| CTR1 | |
| Polyclonal | |
| Unconjugated | |
| SLC31A1 | |
| 4930445G01Rik; AI787263; AU016967; copper transport 1 homolog; copper transporter 1; Copper uptake transporter 1; COPT1; ctr1; ctr-1; hCTR1; high affinity copper uptake protein; high affinity copper uptake protein 1; high-affinity copper uptake protein; liver regeneration-related protein LRRGT00200; LRRGT00200; rCTR1; SLC31A1; slc31a1.L; solute carrier family 13 (sodium/sulphate symporters), member 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporter), member 1 L homeolog; solute carrier family 31 (copper transporters), member 1; solute carrier family 31 member 1; solute carrier family 31, member 1; Xctr1; XELAEV_18038194mg; zgc:76839 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100050369, 100349206, 100689094, 1317, 171135, 20529, 379422, 397186, 403028, 417268, 481678, 518976 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 0.1% sodium azide | |
| O15431, Q8K211, Q8WNR0, Q9JK41 | |
| SLC31A1, slc31a1.L | |
| Synthetic 14 amino acid peptide from C-terminus of human CTR1. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Chicken, Frog, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction