Learn More
CstF-64 Mouse, anti-Human, Clone: 1C5G2, Proteintech
Código de producto. 18030706 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18030706 20 μL 20 microlitros
18077035 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18030706 Proveedor Proteintech N.º de proveedor 685201IG20UL
Mouse Monoclonal Antibody
The CstF-64 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CstF-64. This antibody recognizes human, mouse, rat antigen. The CstF-64 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Antígeno CstF-64
Aplicaciones Western Blot, ELISA
Clon 1C5G2
Concentración 1000 μg/mL
Símbolos de los genes CF 1 64 kDa subunit, CstF 64, CSTF 64 kDa subunit, CSTF2
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 1478
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG1