Learn More
Simport™ Scientific Cryovial™ External Thread with Lip Seal Cryogenic Vials
Designed for storing biological material, human or animal cells, at temperatures as low as -196°C. Simport™ Scientific Cryovial™ External Thread with Lip Seal Cryogenic Vials should be used only in the gas phase of liquid nitrogen.
Especificaciones
| Esterilizables en autoclave | Esterilizable en autoclave |
|---|---|
| Material | Polipropileno |
| Estilo de rosca | Externo |
| Esterilidad | Estéril |
| Color | Natural |
Descripción
- Made of specially formulated polypropylene
- Cap features a long skirt and for one hand aseptic methods
- Fast cap thread design for tightening or removal with a mere 1 1/4 turn and inside thread design will not contribute to contamination
- Lip inside cap ensures leakproof seal even at low temperatures
- Provided with white marking area for sample identification
- Color coded by the use of a CAPINSERT™
- Sterilized by gamma radiation
- Packaged in unique tamperproof, resealable, safety-lock bags
Certifications
CE Marked; RNase, DNase, Pyrogen and DNA-free
Especificaciones
| Esterilizable en autoclave | |
| Externo | |
| Natural |
| Polipropileno | |
| Estéril |
For in vitro diagnostic (IVD) use only.