missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYBB1 Mouse anti-Human, Mouse, Rat, Clone: 6F12B12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60273-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group, none in the acidic group). Beta-crystallins form aggregates of different sizes and are able to self-associate to form dimers or to form heterodimers with other beta-crystallins. This gene, a beta basic group member, undergoes extensive cleavage at its N-terminal extension during lens maturation. It is also a member of a gene cluster with beta-A4, beta-B2, and beta-B3.Especificaciones
| CRYBB1 | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02523, P53674, Q9WVJ5 | |
| CRYBB1 | |
| CRYBB1 Fusion Protein Ag4722 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 6F12B12 | |
| Unconjugated | |
| CRYBB1 | |
| Beta B1 crystallin, Beta crystallin B1, CATCN3, CRYBB1, crystallin, beta B1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 12960, 1414, 25421 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido