missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRTC3 Monoclonal Antibody (R.185.4)

Rabbit Monoclonal Antibody
Marca: Invitrogen™ MA514919
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
It is not recommended to aliquot this antibody.
The protein encoded by this gene has similarity to a yeast protein. It seems to be part of a peripheral membrane protein complex localized on cis/medial Golgi cisternae where it may participate in tethering intra-Golgi transport vesicles.
Especificaciones
| CRTC3 | |
| Monoclonal | |
| 168 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q6UUV7, Q91X84 | |
| Crtc3 | |
| Synthetic peptide corresponding to the sequence of human TORC3 (CRTC3) protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| R.185.4 | |
| Unconjugated | |
| Crtc3 | |
| 2610312F20Rik; 6332415K15Rik; AI429792; BC011210; CREB regulated transcription coactivator 3; CREB-regulated transcription coactivator 3; Crtc3; RGD1309666; TORC3; TORC-3; transducer of CREB protein 3; transducer of regulated cAMP response element-binding protein (CREB) 3; transducer of regulated cAMP response element-binding protein 3; transducer of regulated CREB protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 365297, 64784, 70461 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido