missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRK Recombinant Rabbit Monoclonal Antibody (23GB1405), Invitrogen™

Código de producto. 30087962
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30087962 100 μL 100 microlitros
30087644 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30087962 Proveedor Invitrogen™ N.º de proveedor MA554194

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

The Crk-I and Crk-II forms differ in their biological activities. Crk-II has less transforming activity than Crk-I. Crk- II mediates attachment-induced MAPK8 activation, membrane ruffling and cell motility in a Rac-dependent manner. Involved in phagocytosis of apoptotic cells and cell motility via its interaction with DOCK1 and DOCK4. May regulate the EFNA5-EPHA3 signaling.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CRK
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 23GB1405
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica Crk
N.º de referencia del gen P46108, P47811, P70618, Q16539, Q63768, Q64010
Alias de gen Adapter molecule crk; avian sarcoma virus CT10 (v-crk) oncogene homolog; c-Crk; CRK; CRK proto-oncogene, adaptor protein; CRK1; Crk3; Crk-I; CRKII; Crk-II; CrkIII; Crk-III; Crko; CSAID-binding protein; Csaids binding protein; CSBP; Csbp1; CSBP2; CSPB1; CT-10 related kinase 3; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Exip; Hog; I79_016605; MAP kinase 14; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38 alpha; MAPK 14; MAPK14; MAX-interacting protein 2; mitogen activated protein kinase 14; mitogen-activated protein kinase 14; mitogen-activated protein kinase 14A; mitogen-activated protein kinase p38 alpha; MXI2; p38; p38 alpha; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38a; p38ALPHA; p38-alpha; p38alpha Exip; p38Hog; p38MAPK; PRKM14; PRKM15; Proto-oncogene c-Crk; reactive kinase; RK; SAPK2A; stress-activated protein kinase 2A; tRNA synthetase cofactor p38; v-crk avian sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog (avian); v-crk sarcoma virus CT10 oncogene-like protein
Símbolos de los genes Crk, Mapk14
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human CRKII (1-50AA).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12928, 1398, 1432, 26416, 54245, 81649
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.