missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRHR1 Polyclonal Antibody, Invitrogen™
Descripción
Recommended positive controls: Mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Rabbit (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Corticotropin-releasing factor receptor 1 (CRFR1/CRHR1), a Corticotropin-Releasing Factor Receptor, is involved in mediating the stress response and anxiety-related behaviors. Mutations of the receptor reduce anxiety-related behaviors and may be involved in the pathogenesis of Cushing disease. Multiple isoforms of CRFR1 are produced by alternative splicing. CRFR1 has been reported to be expressed in the pituitary, adrenal gland, brain, colon, ovary, placenta, testis, white blood cells, and skin. ESTs have been isolated from human brain, eye, colon, and embryo libraries.
Especificaciones
Especificaciones
| Antígeno | CRHR1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| génica | CRHR1 |
| N.º de referencia del gen | P34998, P35347 |
| Alias de gen | corticotrophin releasing hormone receptor 1; corticotropin releasing hormone 1; corticotropin releasing hormone receptor 1; corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; corticotropin-releasing hormone receptor 1; CRF 1 receptor; Crf receptor 1; Crf type 1; CRF1; Crf1 receptor; CRFR; CRF-R; CRFR1; CRF-R1; CRF-R-1; CRFR-1; CRF-R1alpha; CRH receptor 1; CRH receptor subtype-1 isoform-3; Crhr; CRH-R 1; CRHR1; CRH-R1; CRH-R-1; CRHR1f; CRH-R1h; CRHR1L; seven transmembrane helix receptor |
| Símbolos de los genes | CRHR1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?