Learn More
CREB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14196-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified.Especificaciones
| CREB5 | |
| Polyclonal | |
| Unconjugated | |
| CREB5 | |
| CRE BPA, CREB 5, CREB5, CREBPA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 231991, 500131, 9586 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02930, Q8K1L0 | |
| Creb5 | |
| CREB5 Fusion Protein Ag5399 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |