missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Creatine Kinase BB Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5110627
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: VISMQKGGNM KEVFTRFCTG LTQIETLFKS KDYEFMWNPH LGYILTCPSN LGTGLRAGVH IKLPNLGKHE KFSEVLKRLR LQKRGTGGVD TAAVGGVFDV SNADRLGFSE VELVQMVVDG VKLLIEMEQR LEQGQAIDDL MPAQK Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 98%.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
Especificaciones
| Creatine Kinase BB | |
| Polyclonal | |
| Unconjugated | |
| CKB | |
| Bck; B-CK; brain creatine kinase; Ck3; Ck-3; CKB; Ckbb; Ckbr; CPK-B; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1152 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P12277 | |
| CKB | |
| Recombinant Protein corresonding to Human Creatine Kinase BB. Recombinant protein control fragment (Product #RP-88888). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido