Learn More
Micronova™ CR100 PC™ Cleanroom Tape—Orange
Código de producto. 12823682 Tienda Micronova Productos
Change view
Click to view available options
Anchura (imperial):
2 in
75 in
Tamaño de la unidad:
cada uno
Código de producto. Anchura (imperial) unitSize
12823682 75 in cada uno
12823672 2 in cada uno
2 options
Código de producto. 12823682
Marca: Micronova™ CR100PC34OR
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Processed for applications in and around cleanrooms
  • Made of vinyl with synthetic rubber adhesive that ensures clean peel from wafer cartridges and most cleanroom work surfaces
  • Tapes and seals curved and irregular surfaces
  • Matte finish can be written on with cleanroom pen
  • Individually packaged in polyethylene bag
Información de órdenes
Available in red, yellow, white, black, brown, dark blue, dark green, light blue, light green, orange, purple, and transparent in 36 yd. long (33m) rolls. Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information on colors not shown here.
Especificaciones
Anchura (imperial) 75 in