missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11609-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a member of the metallo-beta-lactamase family. The encoded protein is a 73kDa subunit of the cleavage and polyadenylation specificity factor and functions as an endonuclease that recognizes the pre-mRNA 3′-cleavage site AAUAAA prior to polyadenylation. It also cleaves after the pre-mRNA sequence ACCCA during histone 3′-end pre-mRNA processing.Especificaciones
| CPSF3 | |
| Polyclonal | |
| Unconjugated | |
| CPSF3 | |
| CPSF, CPSF 73, CPSF 73 kDa subunit, CPSF3, CPSF73, YSH1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 298916, 51692, 54451 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9QXK7, Q9UKF6 | |
| CPSF3 | |
| CPSF3 Fusion Protein Ag2173 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction