Learn More
CPLX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16949-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Positively regulates a late step in synaptic vesicle exocytosis.Especificaciones
| CPLX3 | |
| Polyclonal | |
| Unconjugated | |
| CPLX3 | |
| complexin 3, Complexin III, CPLX3, CPX III, CPXIII, Nbla11589 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 235415, 501005, 594855 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R1B5, Q8WVH0 | |
| CPLX3 | |
| CPLX3 Fusion Protein Ag10571 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |