missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPLX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20249-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is expected to specifically recognize CPLX2 but not other complexins.
Proteins encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release. Two transcript variants encoding the same protein have been found for this gene.Especificaciones
| CPLX2 | |
| Polyclonal | |
| Unconjugated | |
| CPLX2 | |
| 921 L, complexin 2, Complexin II, CPLX2, CPX 2, CPX II, CPX2, Hfb1, Synaphin 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10814, 116657, 12890 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P84086, P84087, Q6PUV4 | |
| CPLX2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido