Learn More
CPEB3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. 16830514 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16830514 20 μL 20 microlitros
16820514 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16830514 Proveedor Proteintech N.º de proveedor 126691AP20UL
Rabbit Polyclonal Antibody
The CPEB3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CPEB3. This antibody recognizes human, mouse, rat antigen. The CPEB3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Antígeno CPEB3
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Polyclonal
Concentración 267 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 22849
Especies diana Human, Rat, Mouse
Isotype IgG