Learn More
Chemsplash CPE OverShoe Compressed Polyethylene -Machine Made
Descripción
Elasticated Overshoe
- 3 GSM Compressed Polyethylene Material
- 16 in. Long
- Elasticated Ankles
- Heat Sealed Seams
- PFAS Free
Description: Elasticated Overshoe in a 3 gram blue compressed polyethylene material. Creates a barrier over your footwear. The 16 in. length offers suitability for all footwear sizing. Ideal for keeping clean footwear clean or keeping clean floors clean from dirty footwear.
Especificaciones
Especificaciones
| Tamaño | 3 g |
| Material | Compressed Polyethylene |
| Latex Free | Yes |
| Recubrimiento | Polyethylene |
| Color | Azul |
| Desechables | Single-use |
| Esterilidad | No estéril |
| Tipo de tapón | Elastic Ankle |
| Tipo de suela | Plana |
| Resistencia | Liquid |
| Mostrar más |