missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX2/ Cyclooxygenase 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Tienda Proteintech ProductosDescripción
PTGS1 (COX1, Cyclooxygenase 1) is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes PTGS1, which regulates angiogenesis in endothelial cells, and is inhibited by nonsteroidal anti-inflammatory drugs such as aspirin. PTGS1 is thought to be involved in cell-cell signaling and maintaining tissue homeostasis. Alternative splicing of this gene generates two transcript variants. The expression of these two transcripts is differentially regulated by relevant cytokines and growth factors.
Especificaciones
Especificaciones
| Antígeno | COX2/ Cyclooxygenase 2 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.23 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | PTGS2 |
| N.º de referencia del gen | P35354, Q05769 |
| Alias de gen | COX 2, COX2, COX2/ Cyclooxygenase 2, Cyclooxygenase 2, GRIPGHS, hCox 2, PGG/HS, PGH synthase 2, PGHS 2, PHS 2, PHS II, Prostaglandin G/H synthase 2, Prostaglandin H2 synthase 2, PTGS2 |
| Símbolos de los genes | PTGS2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?