Learn More
COX10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. p-7088117 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16897093 20 μL 20 microlitros
16887093 150 μL 150 microlitros
2 options
Código de producto. 16897093
Marca: Proteintech 106112AP20UL
Rabbit Polyclonal Antibody
Defects in COX10 are a cause of mitochondrial complex IV deficiency (MT-C4D); also known as cytochrome c oxidase deficiency. A disorder of the mitochondrial respiratory chain with heterogeneous clinical manifestations, ranging from isolated myopathy to severe multisystem disease affecting several tissues and organs. Features include hypertrophic cardiomyopathy, hepatomegaly and liver dysfunction, hypotonia, muscle weakness, excercise intolerance, developmental delay, delayed motor development and mental retardation. A subset of patients manifest Leigh syndrome.
Especificaciones
Antígeno COX10
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.33 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica COX10
N.º de referencia del gen Q12887, Q8CFY5
Alias de gen COX10, Heme O synthase
Símbolos de los genes Cox10
Especie del huésped Rabbit
Inmunógeno COX10 Fusion Protein Ag0930
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1352, 691853, 70383
Especies diana Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG