missing translation for 'onlineSavingsMsg'
Learn More

COX-1/Cyclooxygenase-1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 16891574 Shop All Proteintech Products
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Unit Size:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16891574 150 μL 150 microlitros
16801584 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16891574 Supplier Proteintech Supplier No. 133931AP150UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The COX-1/Cyclooxygenase-1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human COX-1/Cyclooxygenase-1. This antibody recognizes human, mouse, rat antigen. The COX-1/Cyclooxygenase-1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antígeno COX-1/Cyclooxygenase-1
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Polyclonal
Concentración 253 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 150 μL
Primario o secundario Primary
ID de gen (Entrez) 5742
Especies diana Human, Rat, Mouse
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.