Learn More
COX-1/Cyclooxygenase-1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13393-1-AP-20UL
Descripción
The COX-1/Cyclooxygenase-1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human COX-1/Cyclooxygenase-1. This antibody recognizes human, mouse, rat antigen. The COX-1/Cyclooxygenase-1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
| COX-1/Cyclooxygenase-1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 5742 | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 253 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.