Learn More
COTL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16837393 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16837393 20 μL 20 microlitros
16827393 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16837393 Proveedor Proteintech N.º de proveedor 107811AP20UL
Rabbit Polyclonal Antibody
This gene encodes one of the numerous actin-binding proteins which regulate the actin cytoskeleton. This protein binds F-actin, and also interacts with 5-lipoxygenase, which is the first committed enzyme in leukotriene biosynthesis. Although this gene has been reported to map to chromosome 17 in the Smith-Magenis syndrome region, the best alignments for this gene are to chromosome 16. The Smith-Magenis syndrome region is the site of two related pseudogenes.
Especificaciones
Antígeno COTL1
Aplicaciones Immunocytochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Clasificación Polyclonal
Concentración 0.35 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica COTL1
N.º de referencia del gen B0BNA5, Q14019, Q9CQI6
Alias de gen CLP, Coactosin like protein, COTL1
Símbolos de los genes Cotl1
Especie del huésped Rabbit
Inmunógeno COTL1 Fusion Protein Ag1230
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 23406, 361422, 72042
Especies diana Rat, Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG