missing translation for 'onlineSavingsMsg'
Learn More

Anti-coronavirus (SARS-CoV-2) spike S2 (B), mouse, mAb, MP Biomedicals™

Código de producto. p-7087646 Tienda MP Biomedicals Productos
Change view
Click to view available options
Cantidad:
250 μg
50 μg
Tamaño de la unidad:
250 microgramos
50 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16512528 50 μg 50 microgramos
16522528 250 μg 250 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16512528 Proveedor MP Biomedicals N.º de proveedor 08720411

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Monoclonal antibody raised against the coronavirus spike S2 (B) protein, a common target for neutralizing antibodies and vaccines.

Anti-coronavirus (SARS-CoV-2) spike S2 (B), mouse, mAb. The 2019 Novel Coronavirus (SARS-CoV-2) is a coronavirus identified as the cause of an outbreak of respiratory illness first detected in Wuhan, China. Its genome encodes four structural proteins, i.e. spike (S), envelope (E), membrane (M) and nucleocapsid (N). The spike protein (S-protein) contains two subunits, S1 and S2. S1 defines the range of the hosts and specificity of the virus, thus recognizes and binds with the cell surface receptor. S2 subunit contains basic elements needed for the membrane fusion. The spike protein is the common target for neutralizing antibodies and vaccines.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Coronavirus Spike S2 (B) Protein
Aplicaciones Western Blot
Clasificación Monoclonal
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana None
Isotype IgG1

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.