Descripción
Coronin 3 is associated with the aggression and metastasis of hepatocellular carcinoma. Coronins are a family of highly evolutionary conserved proteins reportedly involved in the regulation of actin cytoskeletal dynamics, although only coronin 3 has been shown to be related to cancer cell migration.
Especificaciones
Especificaciones
| Antígeno | CORO1C |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.22 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | CORO1C |
| N.º de referencia del gen | Q9ULV4, Q9WUM4 |
| Alias de gen | CORO1C, Coronin 1C, Coronin 3, CRNN4, HCRNN4 |
| Símbolos de los genes | Coro1c |
| Mostrar más |