Learn More
Corning™ DMEM/Ham's F-12 50/50 Mix, Powder
Used for culture of variety of cell types under low serum conditions. Corning™ DMEM/Ham's F-12 50/50 Mix, Powder ensures improved stability.
Descripción
Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME. Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids. This formulation is without sodium bicarbonate, L-glutamine and phenol red.¿Detecta una oportunidad de mejora?