Learn More
Tocris Bioscience™ CORM 3
Descripción
CORM 3 is a water-soluble carbon monoxide-releasing molecule. Suppresses thrombin-induced nitrite release in BV-2 microglia. Protects isolated rats hearts from reperfusion damage in vitro and is cardioprotective in a rat myocardial infarction model in vivo. Also prolongs survival of transplanted hearts in mice. Exhibits anti-inflammatory and vasorelaxant effects.
Especificaciones
Especificaciones
| Cantidad | 50 mg |
| Tipo de producto | CORM 3 |
| Peso molecular | 294.61 |
Para uso exclusivo en investigación