missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated HVCN1 Mouse Monoclonal Antibody, Proteintech
Tienda Proteintech ProductosDescription
The HVCN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HVCN1. This antibody recognizes human antigen. The HVCN1 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antígeno | HVCN1 |
| Aplicaciones | Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | 1E4C4 |
| Concentración | 1000 μg/mL |
| Conjugado | CoraLite Plus 488 |
| Símbolos de los genes | HV1; MGC15619; VSOP |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant Protein |
| Método de purificación | Protein G purification |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?