missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated ENO1 Mouse Monoclonal Antibody, Proteintech
Tienda Proteintech ProductosDescripción
The ENO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ENO1. This antibody recognizes human, mouse, rat, pig antigen. The ENO1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Especificaciones
Especificaciones
| Antígeno | ENO1 |
| Aplicaciones | Immunocytochemistry/Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | 1D6E8 |
| Concentración | 1000 μg/mL |
| Conjugado | CoraLite Plus 488 |
| Símbolos de los genes | ENO1L1; MBP-1; MPB1; NNE; PPH |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant Protein |
| Método de purificación | Thiophilic affinity chromatograph |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?