missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated EGFR Mouse Monoclonal Antibody, Proteintech
Tienda Proteintech ProductosDescripción
The EGFR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EGFR. This antibody recognizes human antigen. The EGFR antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Especificaciones
Especificaciones
| Antígeno | EGFR |
| Aplicaciones | Flow Cytometry (Intracellular Staining) |
| Clasificación | Monoclonal |
| Clon | 2A2H10 |
| Concentración | 1000 μg/mL |
| Conjugado | CoraLite Plus 488 |
| Símbolos de los genes | ERBB; ERBB1; HER1; PIG61; mENA |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant Protein |
| Método de purificación | Protein G purification |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?