missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated Calbindin-D28k Mouse Monoclonal Antibody, Proteintech

Código de producto. 30236126 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
Código de producto. Cantidad unitSize
30236126 20 μL 20 microlitros
30233361 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 30236126

Brand: Proteintech CL4886639420UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Calbindin-D28k antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Calbindin-D28k. This antibody recognizes human, mouse, rat antigen. The Calbindin-D28k antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.

Specifications

Antígeno Calbindin-D28k
Aplicaciones Immunofluorescence
Clasificación Monoclonal
Clon 1F8B9
Concentración 1000 μg/mL
Conjugado CoraLite Plus 488
Símbolos de los genes CALB
Especie del huésped Mouse
Inmunógeno Recombinant Protein
Método de purificación Protein G purification
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 793
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 28 kDa
Tipo de producto Conjugated Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.