missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Connexin 47 Polyclonal Antibody

Descripción
36-4700 was used in immunofluorescence analysis of Connexin47 (Cx47) on sections of adult mouse brain.
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | Connexin 47 |
| Aplicaciones | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.25 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide; pH 7.4 |
| génica | GJC2 |
| N.º de referencia del gen | Q5T442, Q8BQU6 |
| Alias de gen | B230382L12Rik; connexin 47 type A; connexin 47 type C; connexin 47 type D; connexin-46.6; Connexin-47; CX46.6; Cx47; CXG2; gap junction alpha-12 protein; Gap junction gamma-2 protein; gap junction membrane channel protein alpha 12; gap junction protein gamma 2; gap junction protein, alpha 12; gap junction protein, chi 2; gap junction protein, gamma 2; gap junction protein, gamma 2, 47kDa; GJA12; Gjc2; HLD2; LMPH1C; PMLDAR; SPG44 |
| Símbolos de los genes | GJC2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?