Learn More
Lenz Laborglasinstrumente™ Condensador
Código de producto. 10077720
Click to view available options
N.º de junta cónica estándar:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
Longitud (métrico):
160 mm
250 mm
400 mm
Tamaño de la unidad:
cada uno
Código de producto. 10077720
Marca: Lenz Laborglasinstrumente™ 05210304
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Condensador para vapores de condensación.
  • Resistencia química
  • Resistencia a altas temperaturas
Specifications
Longitud (métrico) 250 mm
N.º de junta cónica estándar NS 29/32
Tipo Condensador
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.